Learn More
Invitrogen™ Human Rabex-5 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1231
Description
RABEX5 Synthetic Peptide, PEP-1231, from Invitrogen.
RABGEF1 forms a complex with rabaptin-5 (RABPT5) that is required for endocytic membrane fusion, and it serves as a specific guanine nucleotide exchange factor (GEF) for RAB5.
Specifications
27342 | |
purified | |
Human Rabex-5 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9UJ41 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
RABGEF1 | |
0.02% sodium azide | |
50 μg | |
PBS |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.