Learn More
Invitrogen™ Human SIRT5 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP1080
Product Code. 15285079
  • 141.00 € / 50µg
Explore more special offers
Description
Description
SIRT5 Synthetic Peptide, PEP-1080, from Invitrogen.
SIRT5 is a human member of a family of proteins called Sirtuins (Sir2-like proteins) and are present in prokaryotes and eukaryotes. All Sir2-like proteins have a sirtuin core domain, which contains a series of sequence motifs conserved in organisms ranging from bacteria to humans. Bacterial, yeast and mammalian sirtuins are able to metabolize NAD and possibly at as mono-ADP-ribosyltransferases. The enzymatic function of sirtuins is not yet completely understood but recent reports of histone-activated Sir2-mediated NAD metabolism and NAD-activated Sir2-mediated histone deacetylation suggest a possible coupled reciprocal activation mechanism involving interactions of Sir2 with NAD and the N epsilon-acetyl-lysine groups of acetylated histones.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
23408
purified
Human SIRT5
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9NXA8
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
SIRT5
0.02% sodium azide
50 μg
PBS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human SIRT5 Synthetic Peptide > 50 μg; Unlabeled