missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Sortilin ELISA Kit
Human Sortilin quantitates human Sortilin in serum, plasma, supernatant.
654.00 € - 4995.00 €
Specifications
Accession Number | 100 kDa NT receptor; 2900053A11Rik; AI852375; glycoprotein 110; glycoprotein 95; Gp110; Gp95; LDLCQ6; mNTR3; neurotensin receptor 3; NT3; NTR3; Nts3; Ntsr3; Sort1; Sortilin; sortilin 1 |
---|---|
Assay Range | 0.819 to 200 ng/mL |
Assay Sensitivity | 0.81 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17135219
|
Invitrogen™
EH433RBX5 |
5 x 96 Tests |
2635.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
17834150
|
Invitrogen™
EH433RBX10 |
10 x 96 Tests |
4995.00 €
Pack of 10 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
17104743
|
Invitrogen™
EH433RB |
96 Tests |
654.00 €
Pack of 96 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
Human Sortilin quantitates human Sortilin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Sortilin.
Sortilin functions as a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Sortilin is required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). It is also required for protein transport from the Golgi apparatus to the endosomes. Sortilin promotes neuronal apoptosis by mediating endocytosis of the proapoptotic precursor forms of BDNF (proBDNF) and NGFB (proNGFB). Also, Sortilin acts as a receptor for neurotensin.Specifications
100 kDa NT receptor; 2900053A11Rik; AI852375; glycoprotein 110; glycoprotein 95; Gp110; Gp95; LDLCQ6; mNTR3; neurotensin receptor 3; NT3; NTR3; Nts3; Ntsr3; Sort1; Sortilin; sortilin 1 | |
0.81 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Sort1 | |
<10% | |
HRP | |
Serum, 50 μL; Supernatant, 50 μL; Plasma, 50 μL | |
Sortilin | |
4 hr 45 min |
0.819 to 200 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q99523 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title