Learn More
Proteintech HumanKine™ recombinant human IL-34 protein
HumanKine™ recombinant protein
Brand:  Proteintech HZ-1316-100UG
 View more versions of this product
Product Code. 17610077
  • 723.00 € / 100µg
Explore more special offers
Description
Description
IL-34 is a 6-α-helix, proinflammatory cytokine that serves as a ligand for colony-stimulating factor-1 receptor (CSF1R). It is expressed most abundantly by spleen cells. It primarily stimulates human peripheral blood monocyte proliferation and macrophage colony formation. It is also required for Langerhans cell and microglia development. IL-34 has been shown to play a protective role in cancer and Alzheimer’s disease, but it can promote the progression of several autoimmune, inflammatory, and metabolic diseases. As such IL-34 could be used as a biomarker to mark the progression of these diseases (PMID: 22579672, 27577879, 31940023).
Specifications
Specifications
Q6ZMJ4
146433
100 μg
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Human
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
>95%
50 mM Acetate pH 4.5 + 75 mM NaCl, See Certificate of Analysis for details
68 and 38 kDa
HEK293
<1 EU/μg
Recombinant
Lyophilized