missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IL-34 protein

HumanKine™ recombinant protein

137.00 € - 3609.00 €

Specifications

Accession Number Q6ZMJ4
Formulation 50 mM Acetate pH 4.5 + 75 mM NaCl, See Certificate of Analysis for details
Gene ID (Entrez) 146433
Molecular Weight (g/mol) 68 and 38 kDa
Source HEK293
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17600077
View Documents Promotion Details
Proteintech
HZ-1316-1000UG
1000 μg
3609.00 €
1000µg
Please to purchase this item. Need a web account? Register with us today!
17610077
View Documents Promotion Details
Proteintech
HZ-1316-100UG
100 μg
723.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
17620077
View Documents Promotion Details
Proteintech
HZ-1316-10UG
10 μg
137.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-34 is a 6-α-helix, proinflammatory cytokine that serves as a ligand for colony-stimulating factor-1 receptor (CSF1R). It is expressed most abundantly by spleen cells. It primarily stimulates human peripheral blood monocyte proliferation and macrophage colony formation. It is also required for Langerhans cell and microglia development. IL-34 has been shown to play a protective role in cancer and Alzheimer’s disease, but it can promote the progression of several autoimmune, inflammatory, and metabolic diseases. As such IL-34 could be used as a biomarker to mark the progression of these diseases (PMID: 22579672, 27577879, 31940023).
Specifications

Specifications

Q6ZMJ4
146433
HEK293
<1 EU/μg
Recombinant
Lyophilized
50 mM Acetate pH 4.5 + 75 mM NaCl, See Certificate of Analysis for details
68 and 38 kDa
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Human
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
>95%
Videos
SDS
Documents

Documents

Product Certifications