missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Calf Serum (New Zealand)
Brand: Cytiva SH30401.02
Code : 84
Additional Details : Weight : 1.00000kg
Description
Supplement cell culture growth with New Zealand-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
Specifications
Albumin, Iron, Lactate Dehydrogenase, Calcium, Chloride, pH, Gamma Globulin, SGOT, IgG, Sodium, TIBC, Alkaline Phosphatase, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
1000 mL | |
0.1 μm filtration | |
≤ 25 EU/mL | |
Yes |
USDA Regulations | |
Calf Serum (New Zealand) | |
Sterile | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes |