missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Calf Serum (New Zealand)
134.00 €
Specifications
Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein |
---|---|
Certifications/Compliance | USDA Regulations |
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Description | Calf Serum (New Zealand) |
Sterility | Sterile |
Description
Supplement cell culture growth with New Zealand-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Sterile | |
≤ 25 EU/mL | |
Yes |
USDA Regulations | |
Calf Serum (New Zealand) | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes |