missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ CDM4Avian Powdered Media
GREENER_CHOICE

Designed for vaccine manufacturing using suspension cell culture with avian cell line.

Brand:  Cytiva SH31035.01

Product Code. 17285896

  • 270.00 € / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

HyClone CDM4Avian cell culture medium is developed for manufacturing of viral vaccines in avian cell lines, such as EB66 cells, and for the production of lytic viruses, such as measles, influenza, and Japanese encephalitis (JE); and of non-lytic viruses, such as the modified vaccinia Ankara (MVA) virus.

  • Chemically defined (CD) animal-derived component-free (ADCF) formulation
  • Designed to support high peak cell density as well as enhanced viral infectivity and productivity
  • Economical, operation-reducing single formulation for cell propagation and viral production
  • Designed for small- to large-scale culture applications
  • Available in liquid and powder formulations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 to 8°C, away from light and moisture
Non-sterile
5 L
Powder
No Glutamine, No Phenol Red, No Sodium Bicarbonate, No Poloxamer 188
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ CDM4Avian Powdered Media > CDM4Avian powder - 5 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.