Learn More
Cytiva HyClone™ CDM4PERMAb cell culture media
Chemically defined Animal-Derived Component-Free (ADCF) media for use with PER.C6 cells
Brand:  Cytiva SH30871.02
Product Code. 16407675
  • 95.90 € / 1 L
Explore more special offers
Description
Description
  • Chemically defined and animal-derived component-free (ADCF) formulation
  • This high-performance cell culture medium has been developed through the Metabolic Pathway Design process to increase process yields in the production of human antibodies and recombinant proteins using PER.C6 technology
  • The medium has been successfully tested in a variety of applications, including fed-batch bioreactor cultures
  • Designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured according to cGMP guidelines
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Liquid
    0.2 μm Sterile-filtered
    Bottle
    Sterile
    1000 mL
    2° to 8°C, away from light
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    One moment while we fetch your results.
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Cytiva HyClone™ CDM4PERMAb cell culture media > Format: Liquid, Quantity: 1000 mL bottle