missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cell Boost™ 7b Supplement
A concentrated amino acid supplement, in a ready to use powder format, for high-yield protein production in CHO fed-batch processes
Brand: Cytiva SH31120.03
505.43 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
- A concentrated solution of amino acids with an alkaline pH
- Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production.
- Designed to support modern manufacturing processes
- Chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
- Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
- Manufactured to meet cGMP manufacturing standards and QC specifications
Specifications
Without: L-glutamine, poloxamer 188, and insulin | |
Amino Acid Supplement | |
HyClone™ |
Liquid | |
High-Yield Protein Production in CHO Fed-Batch Processes | |
1000 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction