missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 7b Supplement

A concentrated amino acid supplement, in a ready to use powder format, for high-yield protein production in CHO fed-batch processes

82.70 € - 534.00 €

Specifications

Description Without: L-glutamine, poloxamer 188, and insulin
Product Type Amino Acid Supplement
For Use With (Application) High-Yield Protein Production in CHO Fed-Batch Processes
Product Line HyClone™
View More Specs

Products 4
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
16574581
View Documents
Cytiva
SH31120.01
100 mL Liquid
82.70 €
100mL
Please to purchase this item. Need a web account? Register with us today!
16584581
View Documents
Cytiva
SH31120.02
500 mL Liquid
345.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
16625805
View Documents
Cytiva
SH31120.03
1000 mL Liquid
534.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
16447695
View Documents
Cytiva
SH31027.01
500 mL Powder
221.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
  • A concentrated solution of amino acids with an alkaline pH
  • Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production.
  • Designed to support modern manufacturing processes
  • Chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
  • Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
  • Manufactured to meet cGMP manufacturing standards and QC specifications
Specifications

Specifications

Without: L-glutamine, poloxamer 188, and insulin
Amino Acid Supplement
High-Yield Protein Production in CHO Fed-Batch Processes
HyClone™
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.