missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Calf Serum, US Origin
Meets the requirements of most users, filtered through triple 100 nm pore size-rated filters.
Brand: Cytiva SH30073.04IH
This item is not returnable.
View return policy
Description
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses

Specifications
Certificate of Suitability: R1-CEP 2000-080 | |
16 to 22 weeks | |
Calf Bovine Serum | |
Irradiated, Heat Inactivated | |
Yes | |
HyClone™ |
Triple filtered through sequential 100 nm pore size-rated filters | |
United States | |
1000 mL | |
<10 EU/mL | |
<10 mg/dL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Characterized Fetal Bovine Calf Serum, US Origin > 1000 mL
Spot an opportunity for improvement?Share a Content Correction