missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)

Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.

Brand:  Cytiva SH30087.03IR25-40

89.06 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 16231102

  • 130.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Sterile
Irradiated, 25 to 40 doses
No
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
500 mL
Sequential-filtered, 100 nm, Gamma-irradiated
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers