missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
24.25 € - 256.00 €
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm, Gamma-irradiated |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
10287963
|
Cytiva
SH30087.02 |
100 mL | None |
24.25 €
100mL |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10136253
|
Cytiva
SH30087.03 |
500 mL | None |
106.00 €
500mL |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10176363
|
Cytiva
SH30087.04 |
1000 mL | None |
206.00 €
1000mL |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12389842
|
Cytiva
SH30087.03HI |
500 mL | Heat Inactivated |
169.00 €
500mL |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16231102
|
Cytiva
SH30087.03IR25-40 |
500 mL | Irradiated, 25 to 40 doses |
126.00 €
500mL |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16241102
|
Cytiva
SH30087.04IR25-40 |
1000 mL | Irradiated, 25 to 40 doses |
224.00 €
1000mL |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16251102
|
Cytiva
SH30087.04IR30-45 |
1000 mL | Irradiated, 30 to 45 doses |
224.00 €
1000mL |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11556541
|
Cytiva
SH30087.04IR |
1000 mL | Gamma Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12369842
|
Cytiva
SH30087.03IR |
500 mL | Gamma Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Age at time of collection: typically 16 - 22 weeks
- Iron-supplemented with additional growth promoting components
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Sterile | |
Sequential-filtered, 100 nm, Gamma-irradiated |