Learn More
Cytiva HyClone™ DPM-PF-CHO Powdered Media
Powder mediums (Main/Base) used to create PF-CHO MPS media
Brand:  Cytiva SH30334.01
 View more versions of this product
Product Code. 17275876
  • 144.00 € / 5 L
Explore more special offers
Description
Description
  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 8°C, away from light and moisture
Powder
No Glutamine, No Phenol Red, No Poloxamer 188
Main powdered media
Non-sterile
5 L