missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ DPM-PF-CHO Powdered Media
Powder mediums (Main/Base) used to create PF-CHO MPS media
141.00 € - 144.00 €
Specifications
Content And Storage | 2 to 8°C, away from light and moisture |
---|---|
Form | Powder |
Sterility | Non-sterile |
Without Additives | No Glutamine, No Phenol Red, No Poloxamer 188 |
Quantity | 5 L |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
17285876
|
Cytiva
SH30335.01 |
Base powdered media |
141.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17275876
|
Cytiva
SH30334.01 |
Main powdered media |
144.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Designed for high cell yield and recombinant protein production
- Allow for direct or sequential adaptation
- Support growth in multiple cell culture systems
- Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
Specifications
2 to 8°C, away from light and moisture | |
Non-sterile | |
5 L |
Powder | |
No Glutamine, No Phenol Red, No Poloxamer 188 |