missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Brand: Cytiva SH30071.03
688.82 EUR valid until 2025-12-31
Use promo code "24268" to get your promotional price.
This item is not returnable.
View return policy
Description
- Enhance growth of cells
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 100 nm pore-size rated filters

Specifications
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
500 mL | |
Sequential Triple-filtered, 100 nm | |
None | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
No |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Fetal Bovine Serum | |
Sterile | |
Characterized | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
HyClone™ | |
USDA Inspected |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.) > 500 mL
Spot an opportunity for improvement?Share a Content Correction