missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Brand: Cytiva SH30070.02IH25-40
This item is not returnable.
View return policy
Description
- Maximize cell culture growth
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 40 nm pore-size rated filters
Specifications
1, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, 25 Dihydroxy Vitamin D, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, Albumin, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Alkaline Phosphatase, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Liquid | |
100 mL | |
Sequential-filtered, 40 nm | |
Heat Inactivated, Irradiated (25 to 40 doses) | |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
No |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
Fetal Bovine Serum | |
Sterile | |
Defined | |
Bluetongue | |
Yes | |
HyClone™ | |
USDA Inspected |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction