missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
134.00 € - 964.00 €
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
Country of Origin | United States |
Product Type | Fetal Bovine Serum |
Sterility | Sterile |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
11586531
|
Cytiva
SH30070.01 |
50 mL | None |
134.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12614217
|
Cytiva
SH30070.01HI |
50 mL | Heat Inactivated |
153.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11591821
|
Cytiva
SH30070.02 |
100 mL | None |
226.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12389782
|
Cytiva
SH30070.02HI |
100 mL | Heat Inactivated |
243.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17266886
|
Cytiva
SH30070.02IH25-40 |
100 mL | Heat Inactivated, Irradiated (25 to 40 doses) | N/A | |||||
17276886
|
Cytiva
SH30070.02IR25-40 |
100 mL | Irradiated (25 to 40 doses) | N/A | |||||
10703464
|
Cytiva
SH30070.03 |
500 mL | None |
938.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772634
|
Cytiva
SH30070.03HI |
500 mL | Heat Inactivated |
964.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12379792
|
Cytiva
SH30070.03IH |
500 mL | Heat Inactivated/Irradiated | N/A | |||||
10369407
|
Cytiva
SH30070.03IR |
500 mL | Irradiated | N/A | |||||
16231082
|
Cytiva
SH30070.03IR25-40 |
500 mL | Irradiated (25 to 40 doses) |
964.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16241082
|
Cytiva
SH30070.03IR30-45 |
500 mL | Irradiated (30 to 45 doses) |
964.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Maximize cell culture growth
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 40 nm pore-size rated filters
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
Sterile | |
Defined | |
≤ 10 mg/dL | |
No |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Fetal Bovine Serum | |
Sequential-filtered, 40 nm | |
≤ 10 EU/mL | |
HyClone™ | |
USDA Inspected |