missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.)

Developed for growth of CHO cells and derivatives, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ II Serum, a fetal bovine serum (FBS) alternative, containing additional growth factors and supplements than basic formulations.

Brand:  Cytiva SH30066.03

 View more versions of this product

Product Code. 10326762

  • 392.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
United States
500 mL
Sequential-filtered, 100 nm
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
Typically less than 2 μg/mL
USDA Inspected
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
FBS Replacement Serum
Sterile
None
≤ 10 EU/mL
≤ 20 mg/dL
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ FetalClone™ II Serum (U.S.) > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.