missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Research Grade Fetal Bovine Serum, South American Origin
Strict manufacturing and quality control procedures ensure quality is suitability for research applications
Brand: Cytiva SV30160.01
This item is not returnable.
View return policy
Description
- Filtered through three sequential 100 nm pore-size rated filters
- Full traceability compliant with current Good Manufacturing Practices (cGMP)
- Extensive quality, performance, and analytical testing
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
- Aseptically packaged in a Class 100 cleanroom
- Pooled after filtration and before final packaging
- Each lot of serum is tested to ensure quality and sterility
- Batch testing available on request
Specifications
Complies with EU Regulations (Directive 92/118/EEC) | |
Fetal Bovine Serum | |
Sterile | |
Research Grade | |
50 mL | |
No | |
Frozen |
South America | |
50 mL | |
Sequential-filtered, 100 nm | |
None | |
Research | |
HyClone™ |