missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Research Grade Fetal Bovine Serum, South American Origin
Strict manufacturing and quality control procedures ensure quality is suitability for research applications
29.13 € - 237.66 €
Specifications
Certifications/Compliance | Complies with EU Regulations (Directive 92/118/EEC) |
---|---|
Country of Origin | South America |
Product Type | Fetal Bovine Serum |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12330273
|
Cytiva
SV30160.02HI |
100 mL |
82.70 €
100mL |
Estimated Shipment: 29-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12336913
|
Cytiva
SV30160.01 |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16635805
|
Cytiva
SV30160.03IR25-40 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Filtered through three sequential 100 nm pore-size rated filters
- Full traceability compliant with current Good Manufacturing Practices (cGMP)
- Extensive quality, performance, and analytical testing
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
- Aseptically packaged in a Class 100 cleanroom
- Pooled after filtration and before final packaging
- Each lot of serum is tested to ensure quality and sterility
- Batch testing available on request
Specifications
Complies with EU Regulations (Directive 92/118/EEC) | |
Fetal Bovine Serum | |
Sequential-filtered, 100 nm | |
Research | |
Frozen |
South America | |
Sterile | |
Research Grade | |
HyClone™ |