Learn More
Cytiva HyClone™ VaccineXpress Liquid Medium
HyClone™ VaccineXpress Liquid Cell Culture Medium is designed for high‐density growth and maintenance of kidney‐derived cell lines (e.g., Vero cells) for viral vaccine development. Well suited for use in multi-well plates and large‐scale manufacturing.
Brand: Cytiva SH31126.01
Description
- VaccineXpress medium is well suited for use in multi-well plates and large‐scale manufacturing using microcarriers in WAVE Bioreactor or Xcellerex bioreactors.
- Serum-free animal-derived component-free formulation.
- Supports direct or sequential adaptation, cell growth and virus propagation.
- Designed to support high peak cell density, viral infectivity, and productivity.
- For use with multiple viral applications (i.e., influenza, rotavirus, RSV, Dengue, Zika).
- Designed for small- to large-scale cell culture applications with use of microcarriers.
- Available in liquid and powder formulations.
- HyClone VaccineXpress medium has been qualified to grow attachment‐dependent Vero cells with microcarriers for producing viral vaccines.
- The lack of both serum and animal‐derived components in the medium reduces variability and enhances scalability and ease of purifying recombinant proteins and viruses in bioprocess applications.
- VaccineXpress is formulated without L‐glutamine for extended shelf life.

Specifications
Store at 2° to 8°C, protected from light. | |
Liquid | |
Serum-free | |
0.2 μm Sterile-filtered | |
No Glutamine, No Poloxamer 199, No Phenol Red | |
1 L |
Animal Origin, Serum-free, Component-free | |
Cell Culture Medium | |
Sterile-filtered | |
Sodium Bicarbonate | |
Designed for small- to large-scale cell culture applications with use of microcarriers. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.