Learn More
HYDRANAL™-CompoSolver E, Honeywell Fluka™
Medium for volumetric one-component Karl Fischer titration (ethanol-based)
Brand:  Honeywell-Hydranal 34734-1L
 View more versions of this product
Product Code. 15624380
  • 97.54 € / 1 L
Explore more special offers
Description
Description
Advantages of Hydranal one-component reagents:
  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HYDRANAL™-CompoSolver E
12°C (53.6°F)
7446-09-5, 693-98-1, 64-17-5
0.80g/mL
1 L
UN1170
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, 2-Methylimidazole, sulphur dioxide