Learn More
HYDRANAL™-CompoSolver E, Honeywell Fluka™
Medium for volumetric one-component Karl Fischer titration (ethanol-based)
97.54 € - 640.16 €
Specifications
Density | 0.80g/mL |
---|---|
Flash Point | 12°C (53.6°F) |
Chemical Name or Material | HYDRANAL™-CompoSolver E |
CAS | 7446-09-5, 693-98-1, 64-17-5 |
UN Number | UN1170 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15624380
|
Honeywell-Hydranal
34734-1L |
1 L |
97.54 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15634380
|
Honeywell-Hydranal
34734-2.5L |
2.5 L |
183.24 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654380
|
Honeywell-Hydranal
34734-6X1L |
6 x 1 L |
509.29 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15644380
|
Honeywell-Hydranal
34734-4X2.5L |
4 x 2.5 L |
640.16 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
Specifications
0.80g/mL | |
HYDRANAL™-CompoSolver E | |
UN1170 |
12°C (53.6°F) | |
7446-09-5, 693-98-1, 64-17-5 |
Safety and Handling
P201-P210-P280-P308 + P313-P370 + P378-P403 + P235
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethanol; ethyl alcohol, 2-Methylimidazole, sulphur dioxide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.