Learn More
HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™
Brand:  Honeywell-Hydranal 34836-500ML
130.19 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15624500
  • 137.68 € / 500mL
Explore more special offers
Specifications
HYDRANAL™-Coulomat AG
500 mL
UN1230
14°C (57.2°F)
67-56-1, 111-42-2, 7446-09-5, 288-32-4, 68007-08-9
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™ >