Learn More
HYDRANAL™-Moisture Test Kit, Honeywell Fluka™
Test kit for the visual water determination according to Karl Fischer without titrator
Brand:  Honeywell-Hydranal 37858-1EA
Product Code. 15685160
  • 342.75 € / Each
Explore more special offers
Description
Description
Advantages of HYDRANAL- Auxiliaries
  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HYDRANAL™-Moisture Test Kit
12°C (53.6°F)
1 Pc.
UN1993BE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
HYDRANAL™-Moisture Test Kit, Honeywell Fluka™ > 1EA KIT