Learn More
HYDRANAL™ - Titrant 5, Honeywell Fluka™
Titrating agent for volumetric, two-component Karl Fischer titrations (Methanol-based)
Brand: Honeywell Fluka 34801-1L-OM
View more versions of this product
Description
Advantages of Hydranal™ Two-Component Reagents:
- High titration speed
- Ideal accuracy for small amounts of water
- High buffer capacity
- Exact and stable titer
- E-type reagents offer reduced toxicity compared to methanol

Specifications
HYDRANAL™ - Titrant 5 | |
0.850 g/cm3 (at 20°C) | |
1 L | |
Neutral | |
Metrohm OMNIS Smart Reagent | |
Liquid | |
Methyl Alcohol | |
Volumetric Two-Component KF Titration |
Brown | |
11°C | |
7553-56-2 | |
65°C (at 1.013 hPa) | |
-98°C | |
UN1230 | |
Completely Miscible in Water |
Safety and Handling
- HYDRANAL NEXTGEN Titrant 5 OM
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, iodine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.