missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, 48%, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Specifications
pH | lt;1 |
Color | Yellow |
Melting Point | -11°C |
Boiling Point | 126°C |
Packaging | HDPE plastic bottle |
Vapor Pressure | 8mm Hg 25 |
Quantity | 2.5 L |
Physical Form | Liquid |
Formula Weight | 80.9g/mol |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling


Product Identifier
- Hydrobromic acid, ca. 48 wt% solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrobromic Acid, 48%, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction