Learn More
Hydrobromic acid, 48% w/w aq. soln., Thermo Scientific Chemicals
Description
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 1.49 g/mL |
Sensitivity | Air Sensitive |
Quantity | 940 mL |
Physical Form | Liquid |
Beilstein | 3587158 |
Solubility Information | Miscible with water,alcohol and organic solvents. |
Formula Weight | 80.92 |
Chemical Name or Material | Hydrobromic acid |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M28L043 | Certificate of Analysis | 30/01/2025 | 014036.AT,014036.AZ |
U20K053 | Certificate of Analysis | 06/09/2024 | 014036.AT,014036.AZ |
10240267 | Certificate of Analysis | 05/02/2024 | 014036.AT,014036.AZ |
T05J034 | Certificate of Analysis | 13/07/2023 | 014036,014036.AT,014036.AZ |
Q27J012 | Certificate of Analysis | 05/05/2023 | 014036.AT |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.