missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.1 N standard solution, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 124200100

106.65 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15294614

  • 149.00 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hydrochloric Acid
0.1 N Standard Solution
100.0°C
1.0000g/mL
1
ClH
Liquid
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
Colorless
7647-01-0, 7732-18-5
0.0998 to 0.1002N (20°C)
Plastic Drum
10 L
HCl
MFCD00011324 MFCD00792839
15, 4818
Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene
Cl
36.46
CHEBI:17883
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only