missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid 36 - 38%, BAKER ANALYZED™, J.T. Baker™
Laboratory Reagent
39.00 € - 67.00 €
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10381701
|
JT Baker
6081.1000 |
1 L |
39.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10148050
|
JT Baker
6081.2500 |
2.5 L |
67.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrogen chloride |
Specifications
7647-01-0 | |
110°C (1013 hPa) | |
max. 5 ppm | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride |
-30°C | |
1.18 g/cm³ (20°C) | |
37 to 38% | |
MFCD00011324 MFCD00792839 | |
Cl | |
36.46 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrochloric acid 25-38%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title