missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
21.40 € - 180.00 €
Chemical Identifiers
CAS | 7647-01-0, 7732-18-5 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10790872
|
Thermo Scientific Acros
423795000 |
500 mL | Glass Bottle |
21.60 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10270593
|
Thermo Scientific Acros
423795001 |
500 mL | Plastic bottle |
21.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10167560
|
Thermo Scientific Acros
423790025 |
2.5 L | EcoSafPak™ Glass Bottle |
55.10 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10117832
|
Thermo Scientific Acros
423790026 |
2.5 L | Plastic bottle |
53.90 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10699283
|
Thermo Scientific Acros
423790251 |
25 L | Plastic drum |
180.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-01-0, 7732-18-5 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
7647-01-0, 7732-18-5 | |
1ppm max. | |
ClH | |
Solution, Free From Suspended Matter or Sediment | |
1789 | |
15, 4818 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.45 | |
ACS Reagent |
1.1800g/mL | |
0.0005% max. | |
HCl | |
MFCD00011324 MFCD00792839 | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
36.5 to 38.0% | |
Hydrochloric acid, ca. 37% Solution in Water |
Safety and Handling
May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage
Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrochloric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P234-Keep only in original container.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only