Learn More
Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -35.0°C |
Boiling Point | 57.0°C |
Density | 1.1800g/mL |
Heavy Metals (as Pb) | 2ppm max. |
Packaging | Plastic bottle |
Reducing Substances | (SO2) Passes Test |
Specific Gravity | 1.18 |
Quantity | 2.5 L |
Viscosity | 1.9 mPa.s (15°C) |
Linear Formula | HCl |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2400126 | Certificate of Analysis | 31/01/2025 | 124630025,124630011,124630100,124630026,124635001,124630010,124635000 |
2494581 | Certificate of Analysis | 31/01/2025 | 124630025,124630011,124630100,124630026,124635001,124630010,124635000 |
2415249 | Certificate of Analysis | 31/01/2025 | 124630025,124630011,124630100,124630026,124635001,124630010,124635000 |
2413699 | Certificate of Analysis | 31/01/2025 | 124630025,124630011,124630100,124630026,124635001,124630010,124635000 |
2416863 | Certificate of Analysis | 31/01/2025 | 124630025,124630011,124630100,124630026,124635001,124630010,124635000 |
Safety and Handling


- Hydrochloric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.