missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals Product Code.: 10133080

Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals

Product Code. 10133080
500 mL, Glass Bottle
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10133080

Brand: Thermo Scientific Acros 124635000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstofShow MoreShow Less
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Melting Point -35.0°C
Boiling Point 57.0°C
Density 1.1800g/mL
Heavy Metals (as Pb) 2ppm max.
Packaging Glass Bottle
Reducing Substances (SO2) Passes Test
Specific Gravity 1.18
Quantity 500 mL
Viscosity 1.9 mPa.s (15°C)
Linear Formula HCl
Physical Form Solution
Fieser 04,450; 05,533; 06,283
Solubility Information Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
IUPAC Name chlorane
Formula Weight 36.46
Percent Purity 35.0 to 39.0%
Grade Analytical
Chemical Name or Material Hydrochloric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2400126 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124630025,124630011,124630100,124630026,124635001,124630010,124635000
Lot Number2494581 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124630025,124630011,124630100,124630026,124635001,124630010,124635000
Lot Number2415249 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124630025,124630011,124630100,124630026,124635001,124630010,124635000
Lot Number2413699 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124630025,124630011,124630100,124630026,124635001,124630010,124635000
Lot Number2416863 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124630025,124630011,124630100,124630026,124635001,124630010,124635000
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.