missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof, acide chlorhydrique, chlorohydric acid, muriatic acid, chlorwasserstoff, hydrochloric acid, 6AMP, CHEBI:17883, Colorless to Yellow, VEXZGXHMUGYJMC-UHFFFAOYSA-N
21.10 € - 77.80 €
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10590721
|
Fisher Chemical
J/4330C/05 |
1 Ampule |
21.10 €
1 ampoule |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10315710
|
Fisher Chemical
J/4330C/90 |
6 Ampules |
77.80 €
6 ampoules |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof, acide chlorhydrique, chlorohydric acid, muriatic acid, chlorwasserstoff, hydrochloric acid, 6AMP, CHEBI:17883, Colorless to Yellow, VEXZGXHMUGYJMC-UHFFFAOYSA-N
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
lt; 1 | |
7647-01-0 | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 |
Colorless to Yellow | |
Ampoule | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 0.5M (0.5N) Hydrochloric acid (5M Concentrate for volumetric analysis)
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title