missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 0.05 M (0.05 N) Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Fully factorized, high purity, stable, certified product, developed and tested for acid/base titrations.

Brand:  Reagecon HF20051

Product Code. 11576783

  • 32.20 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Acid/base titrations, also called neutralization titrations, are used to determine either the amount of acid/base in a sample or the substance that can be converted to an acid/base.
Specifications

Specifications

0.0 5M (0.05 N)
HDPE Bottle GL45
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberHF200525A1 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog NumberHF20051
Lot NumberHF200524B1 Certificate TypeCertificate of Analysis Date01/12/2024 Catalog NumberHF20051
Lot NumberHF200522J1 Certificate TypeCertificate of Analysis Date20/12/2022 Catalog NumberHF20051
Lot NumberHF200522F1 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog NumberHF20051
Lot NumberHF200521E1 Certificate TypeCertificate of Analysis Date02/11/2021 Catalog NumberHF20051
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrofluoric Acid 0.05 M (0.05 N) Analytical Volumetric Solution (AVL), Reagecon > 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.