Learn More
Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals
34.40 € - 44.50 €
Chemical Identifiers
CAS | 7664-39-3, 7732-18-5 |
---|---|
Molecular Formula | FH |
Molecular Weight (g/mol) | 20.01 |
MDL Number | MFCD00011346 |
InChI Key | KRHYYFGTRYWZRS-UHFFFAOYSA-N |
Synonym | hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 |
PubChem CID | 14917 |
ChEBI | CHEBI:29228 |
IUPAC Name | hydrogen fluoride |
SMILES | F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10637581
|
Thermo Scientific Acros
|
25 g | Plastic bottle |
34.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10741452
|
Thermo Scientific Acros
423805000 |
500 g | Plastic bottle |
44.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7664-39-3, 7732-18-5 | |
20.01 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
14917 | |
hydrogen fluoride |
FH | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
CHEBI:29228 | |
F |
Specifications
Colorless | |
-35.0°C | |
1.1500g/mL | |
0.0005% max. | |
FH | |
Liquid | |
06,284; 09,238; 16,176 | |
Solubility in water: soluble. | |
F | |
20.01 | |
CHEBI:29228 | |
48.0 to 51.0% | |
Hydrofluoric acid, 48.0-51.0% |
7664-39-3, 7732-18-5 | |
105.0°C | |
0.5ppm max. | |
1.15 | |
HF | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
hydrogen fluoride | |
14917 | |
20 | |
ACS Reagent |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal if inhaled.
GHS P Statement:
Wear face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 231-634-8
missing translation for 'rtecsNumber' : MW7875000
missing translation for 'tsca' : TSCA
- Hydrofluoric acid, 48 to 51% solution in water
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H310-Fatal in contact with skin.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H300-Fatal if swallowed.
- H330-Fatal if inhaled.
- H290-May be corrosive to metals.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only