missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥48%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Concentration | ≥48% |
Density | 1.15g/mL at 25°C |
Residue after Ignition | ≤0.0005% (as SO4) |
Vapor Pressure | 25mmHg (20°C) |
Quantity | 1 L |
UN Number | 1790 |
IUPAC Name | fluorane |
Formula Weight | 20.01g/mol |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Hydrofluoric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling


Product Identifier
- Hydrofluoric acid
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrogen fluoride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrofluoric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥48%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction