missing translation for 'onlineSavingsMsg'
Learn More

Contec™ HydroPure™ with Purified Water
BIOPHARMA

HydroPure™ with Purified Water designed for cleaning of life science cleanrooms.

Brand:  Contec™ SBT16HPW

178.03 EUR valid until 2024-12-20
Use promo code "21731" to get your promotional price.



 View more versions of this product

Product Code. 15567171

  • 246.00 € / 6 bottles
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed for cleaning of life science cleanrooms.

  • Composition: 6% hydrogen peroxide in water
  • Can be applied directly to surface ensuring even coverage
  • Filtered to 0.22 microns under Grade A airflow
  • Physically removes contaminants
  • Sufficient sporicidal activity for Grade A / B area
  • Low residue, eliminates need for residue removal stage with alcohol or water
  • Gamma irradiated
  • Large droplet size
  • Facilitates transfer disinfection into isolators and RABS
  • Same grade as when vaporized hydrogen peroxide is used
  • Triple / double bagged in linear tear packaging
  • Noncorrosive
Applications:
  • Efficacious against bacteria, fungi, moulds, yeasts and spores
Specifications

Specifications

Detergent with Purified Water
6.5 to 7.5
Colorless
Clear
6% Hydrogen peroxide in purified water (EP)
Limited-use
No
6 x 1 L
1 L
Liquid Spray
Sterile
Not Autoclavable
Hydrogen peroxide and purified water (EP)
Trigger Spray Bottle
Filtered
Used in pharmaceutical industry
33.81 oz.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CONTEC STERILE HYDROPURE

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : HYDROGEN PEROXIDE SOLUTION

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.