missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ HydroPure™ with Purified Water

HydroPure™ with Purified Water designed for cleaning of life science cleanrooms.
231.00 € - 246.00 €
Specifications
Product Type | Detergent with Purified Water |
---|---|
pH | 6.5 to 7.5 |
Sterility | Sterile |
Appearance | Colorless |
Autoclavable | Not Autoclavable |
Product Code | Brand | Container Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Container Type | Quantity | Price | Quantity & Availability | ||||
15517171
![]() |
Contec™
SBC56HP |
Capped Bottle | 2 x 5 L |
231.00 €
2 bottles |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15567171
![]() |
Contec™
SBT16HPW |
Trigger Spray Bottle | 6 x 1 L |
246.00 €
6 bottles |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Designed for cleaning of life science cleanrooms.
- Composition: 6% hydrogen peroxide in water
- Can be applied directly to surface ensuring even coverage
- Filtered to 0.22 microns under Grade A airflow
- Physically removes contaminants
- Sufficient sporicidal activity for Grade A / B area
- Low residue, eliminates need for residue removal stage with alcohol or water
- Gamma irradiated
- Large droplet size
- Facilitates transfer disinfection into isolators and RABS
- Same grade as when vaporized hydrogen peroxide is used
- Triple / double bagged in linear tear packaging
- Noncorrosive
- Efficacious against bacteria, fungi, moulds, yeasts and spores
Specifications
Detergent with Purified Water | |
Sterile | |
Not Autoclavable | |
Hydrogen peroxide and purified water (EP) | |
Filtered | |
Used in pharmaceutical industry |
6.5 to 7.5 | |
Colorless | |
Clear | |
Limited-use | |
No |
Safety and Handling
missing translation for 'shelfLife' : 1 Year
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CONTEC STERILE HYDROPURE
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : HYDROGEN PEROXIDE SOLUTION
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Contec™ HydroPure™ with Purified Water