missing translation for 'onlineSavingsMsg'
Learn More
Ansell Edmont™ Hyflex™ 11-518 Series Ultra Light Duty Polyurethane Gloves Product Code.: 12618665
Add to basket

Ansell Edmont™ Hyflex™ 11-518 Series Ultra Light Duty Polyurethane Gloves

Product Code. 12618665 Shop All Ansell Products
Size: 7
Click to view available options
Size: 7
This item is not returnable. View return policy

Product Code. 12618665

Brand: Ansell Edmont™ 11518070

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Tailored to hand fit, creates the sensation of bare-hand comfort. Ansell Edmont™ Hyflex™ 11-518 Series Ultra Light Duty Polyurethane Gloves are ultra light duty cut resistant gloves.

  • Industrial gloves
  • Protects workers with this first truly Lightweight glove with EN Level 3 cut protection
  • Soft ultra light-duty seamless liner breathes to keep hands cool and dry, and offers enhanced flexibility
  • Good fingertip sensitivity with superior comfort
  • Dyneema™ Diamond Technology fiber thin PU coating bring unmatched dexterity and tactility for applications with cut risks

Applications:

  • Applying finish to materials, products
  • Assembly & inspection of components
  • Delivery of parts to production line
  • Final assembly
  • Handling incoming goods
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Picking, fastening components
  • Press injection molding
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2006
Description Gauge: 18
Color Blue
Exterior Finish Palm Coated
Material Polyurethane
Product Type Industrial Gloves
Size 7
Style Construction: Knitted coated
Glove Cuff Style Knit
Liner Material Nylon - Spandex - Dyneema™ Diamond Technology
Packaging Type 12 pairs in a bag, 12 bags in a carton
Series 11-518
Length (Metric) 210 to 265 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Edmont™ Hyflex™ 11-518 Series Ultra Light Duty Polyurethane Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.