missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates

Use Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates with your HyperSep system.

Brand:  Thermo Scientific™ 60300-403

687.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10595225

  • 690.56 € / Pack of 50
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates with your HyperSep system.

Fast analysis without sacrificing accuracy or reproducibility. Incorporate traditional solid phase extraction sorbents into a 96-well plate format using the HyperSep system.

  • Innovative design of 96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation
  • Well plates are customizable to allow any arrangement of wells with differing sorbent chemistries for simultaneous processing.


Ordering Information:
Available in packs of 100 individual wells or in pre-assembled plates.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample Collection Plate, 2 mL
Vacuum Manifold
Sample Collection Plate
SPE Sample Preparation
HyperSep
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Sample Collection Plates > Sample collection plate, 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.