Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Manifold
Use this flexible vacuum manifold to process samples using both solid phase extraction (SPE) cartridges and 96 well plates.
Brand:  Thermo Scientific™ 60104-230
2671.33 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 10641704
  • 2819.10 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Process your samples in solid phase extraction SPE cartridges or 96 well plates by using the flexible Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. The manifold is supplied with a base and vacuum gauge, flask and stopper tubing and spigots.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal Vacuum Manifold, 1 pk
SPE Columns and 96 Well Plates
Universal Vacuum Manifold
SPE
HyperSep
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Universal Vacuum Manifold > For HyperSep SPE columns and HyperSep-96 well plates
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.