missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ BDS C18 Column, 3μm

For a robust, general-purpose column to use in a wide range of applications, choose Thermo Scientific™ Hypersil™ BDS C18 HPLC Columns.

Brand:  Thermo Scientific™ 28103-124030

 View more versions of this product

Product Code. 10252488

  • 802.86 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Base-deactivated with minimal residual silanol activity
  • Exceptional reproducibility
  • Economical, general purpose columns
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
125 mm
2 to 9
170 m²/g
C18
11%
Reversed Phase
4 mm
3 μm
130 Å
L1
Silica
Reversed Phase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers