Learn More
Thermo Scientific™ Hypersil GOLD™ PREP C18 HPLC Columns
Easily scale up your method to prep LC using Hypersil GOLD C18 preparative HPLC columns for excellent peak shape in a variety of formats including XtendedLife technology.
246.90 € - 28591.02 €
Specifications
pH 2 to 9
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
USP Type L1
View More Specs
Products 34
Product Code Brand Length (Metric) Diameter (Metric) Column Format Particle Size Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Column Format Particle Size Price Quantity & Availability  
16378506
View Documents
Thermo Scientific™
25005-059070A
50 mm 10 mm Preparative Column, Standard 5 μm
1363.23 €
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
16388506
View Documents
Thermo Scientific™
25005-059270A
50 mm 20 mm Preparative Column, Standard 5 μm
2916.43 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16398506
View Documents
Thermo Scientific™
25005-059270XL
50 mm 25 mm Preparative Column, Xtended Life 5 μm
7507.41 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
16308516
View Documents
Thermo Scientific™
25005-109070A
100 mm 10 mm Preparative Column, Standard 5 μm
1797.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16318516
View Documents
Thermo Scientific™
25005-109270A
100 mm 20 mm Preparative Column, Standard 5 μm
3807.40 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
16328516
View Documents
Thermo Scientific™
25005-109270XL
100 mm 25 mm Preparative Column, Xtended Life 5 μm
7158.43 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16338516
View Documents
Thermo Scientific™
25005-159070A
150 mm 10 mm Preparative Column, Standard 5 μm
2175.53 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
16348516
View Documents
Thermo Scientific™
25005-159270A
150 mm 20 mm Preparative Column, Standard 5 μm
5086.58 €
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
16358516
View Documents
Thermo Scientific™
25005-159270XL
150 mm 25 mm Preparative Column, Xtended Life 5 μm
7945.08 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16368516
View Documents
Thermo Scientific™
25005-159370A
150 mm 30 mm Preparative Column, Standard 5 μm
8715.60 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16378516
View Documents
Thermo Scientific™
25005-159370XL
150 mm 30 mm Preparative Column, Xtended Life 5 μm N/A N/A N/A
16388516
View Documents
Thermo Scientific™
25005-259070A
250 mm 10 mm Preparative Column, Standard 5 μm
2354.70 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16398516
View Documents
Thermo Scientific™
25005-259270A
250 mm 20 mm Preparative Column, Standard 5 μm
5695.46 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
16308526
View Documents
Thermo Scientific™
25005-259270XL
250 mm 25 mm Preparative Column, Xtended Life 5 μm
9331.59 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16318526
View Documents
Thermo Scientific™
25005-259370A
250 mm 30 mm Preparative Column, Standard 5 μm
11579.49 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16328526
View Documents
Thermo Scientific™
25005-259370XL
250 mm 30 mm Preparative Column, Xtended Life 5 μm N/A N/A N/A
16338526
View Documents
Thermo Scientific™
25012-018823A
8 mm 10 mm Guard Column 12 μm
246.90 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
16348526
View Documents
Thermo Scientific™
25012-019023A
20 mm 10 mm Guard Column 12 μm
368.25 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
16358526
View Documents
Thermo Scientific™
25012-019323A
30 mm 10 mm Guard Column, 1/16 in. 12 μm
519.22 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16368526
View Documents
Thermo Scientific™
25012-019323B
30 mm 10 mm Guard Column, 1/8 in. 12 μm
519.22 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16378526
View Documents
Thermo Scientific™
25012-029023A
20 mm 20 mm Guard Column 12 μm N/A N/A N/A
16388526
View Documents
Thermo Scientific™
25012-029323A
30 mm 20 mm Guard Column, 1/16 in. 12 μm
774.60 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16398526
View Documents
Thermo Scientific™
25012-029323B
30 mm 20 mm Guard Column, 1/8 in. 12 μm
774.60 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16308536
View Documents
Thermo Scientific™
25012-159270A
150 mm 20 mm Preparative Column, Standard 12 μm
3950.98 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16318536
View Documents
Thermo Scientific™
25012-159270XL
150 mm 25 mm Preparative Column, Xtended Life 12 μm
7738.06 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16328536
View Documents
Thermo Scientific™
25012-159370A
150 mm 30 mm Preparative Column, Standard 12 μm
8330.98 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16338536
View Documents
Thermo Scientific™
25012-159370XL
150 mm 30 mm Preparative Column, Xtended Life 12 μm N/A N/A N/A
16348536
View Documents
Thermo Scientific™
25012-259070A
250 mm 10 mm Preparative Column, Standard 12 μm
2072.49 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16358536
View Documents
Thermo Scientific™
25012-259270A
250 mm 20 mm Preparative Column, Standard 12 μm
5401.79 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
16368536
View Documents
Thermo Scientific™
25012-259270XL
250 mm 25 mm Preparative Column, Xtended Life 12 μm
9011.22 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16378536
View Documents
Thermo Scientific™
25012-259370A
250 mm 30 mm Preparative Column, Standard 12 μm
11167.30 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16388536
View Documents
Thermo Scientific™
25012-259370XL
250 mm 30 mm Preparative Column, Xtended Life 12 μm N/A N/A N/A
16398536
View Documents
Thermo Scientific™
25012-259570A
250 mm 50 mm Preparative Column, Standard 12 μm
28591.02 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
16308546
View Documents
Thermo Scientific™
25012-259570XL
250 mm 50 mm Preparative Column, Xtended Life 12 μm N/A N/A N/A
Description
Show More
Specifications
Specifications
2 to 9
220 m²/g
L1
Spherical, Fully Porous, Ultrapure Silica
Reversed Phase
Reversed Phase
175 Å
60°C
C18
10%
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Hypersil GOLD™ PREP C18 HPLC Columns