missing translation for 'onlineSavingsMsg'
Learn More

Corning™ HYPERStack™ Untreated Cell Culture Vessels - New Design
BIOPHARMAGREENER_CHOICE

More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint.

Brand:  Corning™ 20013

 View more versions of this product

Product Code. 17203084

  • 3385.00 € / Pack of 4
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Descrizione

Descrizione

Corning's High Yield PERformance (HYPER) Platform - Closed System for High Yield Cell Growth

The Corning HYPERStack cell culture vessel combines the best of two Corning products: the Corning CellSTACK™ and the Corning HYPERFlask™ cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be among the most efficient, scalable cell culture vessels for adherent cell culture available today.

Features and Benefits

  • More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
  • Closed system – no open fluid manipulations
  • Scalable product – multiple size offerings support scale-up and scale-out
  • Ergonomic design – easier manipulation with handling equipment and accessories
  • Fixed media volume – 0.2 mL/cm2 fills vessel for less volumetric waste

Surface Treatment Guide

Corning CellBIND™ surface treatment is designed to enhance cell attachment and increase cell yields, improving attachment of loosely adherent cells and maintaining growth in reduced-serum or serum-free conditions. Alternatively, a non-treated option can support conventional cell attachment and allow for a faster and gentler harvest procedure for downstream processing and assays.

TRUSTED_SUSTAINABILITY
Specifica

Specifica

1310 mL
4/Cs.
12-layer vessel
Sterile
Non-treated
Polystyrene
1 Ea.
Rectangular
6000 cm2
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali