Learn More
Corning™ HYPERStack™ Untreated Cell Culture Vessels - New Design
More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint.
3325.00 € - 3385.00 €
Specifications
Material | Polystyrene |
---|---|
No. per Pack | 1 Ea. |
Shape | Rectangular |
Sterility | Sterile |
Surface Treatment | Non-treated |
Product Code | Brand | Culture Area | Product Type | No. per Case | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Product Type | No. per Case | Price | Quantity & Availability | |||
17203084
|
Corning™
20013 |
6000 cm2 | 12-layer vessel | 4/Cs. |
3385.00 €
Pack of 4 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
|||
17223084
|
Corning™
20037 |
18,000 cm2 | 36-layer vessel | 2/Cs. |
3325.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
Description
Corning's High Yield PERformance (HYPER) Platform - Closed System for High Yield Cell Growth
The Corning HYPERStack cell culture vessel combines the best of two Corning products: the Corning CellSTACK™ and the Corning HYPERFlask™ cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be among the most efficient, scalable cell culture vessels for adherent cell culture available today.
Features and Benefits
- More cells – provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
- Closed system – no open fluid manipulations
- Scalable product – multiple size offerings support scale-up and scale-out
- Ergonomic design – easier manipulation with handling equipment and accessories
- Fixed media volume – 0.2 mL/cm2 fills vessel for less volumetric waste
Surface Treatment Guide
Corning CellBIND™ surface treatment is designed to enhance cell attachment and increase cell yields, improving attachment of loosely adherent cells and maintaining growth in reduced-serum or serum-free conditions. Alternatively, a non-treated option can support conventional cell attachment and allow for a faster and gentler harvest procedure for downstream processing and assays.
Specifications
Polystyrene | |
Rectangular | |
Non-treated |
1 Ea. | |
Sterile |