Learn More
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Make procedures as painless as possible with extremely sharp and reliable stainless steel needles. Terumo™ Agani™ Single-use Sterile Hypodermic Needles feature a unique, latex-free double bevel design for maximum comfort.
7.80 € - 14.05 €
Specifications
Product Type | Sterile Hypodermic Needle for Single-Use |
---|---|
Sterility | Sterile |
Product Code | Brand | Needle Gauge | Color | Needle Point Style | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Color | Needle Point Style | Length (Metric) | Price | Quantity & Availability | ||
15351557
|
Terumo™
8AN1838S1 |
18 G | Pink | 17° Short | 38 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15331537
|
Terumo™
8AN1925R1 |
19 G | Ivory | 11° Regular | 25 mm |
8.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15341537
|
Terumo™
8AN1938R1 |
19 G | Ivory | 11° Regular | 38 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15361557
|
Terumo™
8AN1938S1 |
19 G | Ivory | 17° Short | 38 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15428652
|
Terumo™
8AN1950R1 |
19 G | Ivory | 11° Regular | 50 mm |
8.55 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15371537
|
Terumo™
8AN2116R1 |
21 G | Green | 11° Regular | 16 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15438652
|
Terumo™
8AN2150R1 |
21 G | Green | 11° Regular | 50 mm |
8.85 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15301547
|
Terumo™
8AN2232R1 |
22 G | Black | 11° Regular | 32 mm |
8.25 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15331547
|
Terumo™
8AN2316R1 |
23 G | Blue | 11° Regular | 16 mm |
8.20 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15351547
|
Terumo™
8AN2332R1 |
23 G | Blue | 11° Regular | 32 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15361547
|
Terumo™
8AN2425R1 |
24 G | Purple | 11° Regular | 25 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15391547
|
Terumo™
8AN2525R1 |
25 G | Orange | 11° Regular | 25 mm |
8.10 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15331557
|
Terumo™
8AN2716R1 |
27 G | Gray | 11° Regular | 16 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15391557
|
Terumo™
8AN3013R1 |
30 G | Yellow | 11° Regular | 13 mm |
14.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15311557
|
Terumo™
8AN2623R1 |
26 G | Brown | 11° Regular | 23 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15301557
|
Terumo™
8AN2613R1 |
26 G | Maroon | 11° Triple Bevel | 13 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15371547
|
Terumo™
8AN2516R1 |
25 G | Orange | 11° Triple Bevel | 16 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15361537
|
Terumo™
8AN2038R1 |
20 G | Yellow | 11° Triple Bevel | 38 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15321547
|
Terumo™
8AN2238R1 |
22 G | Black | 11° Triple Bevel | 38 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15341547
|
Terumo™
8AN2325R1 |
23 G | Blue | 11° Triple Bevel | 25 mm |
7.80 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15341557
|
Terumo™
8AN2719R1 |
27 G | Gray | 11° Triple Bevel | 19 mm |
8.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15391537
|
Terumo™
8AN2138R1 |
21 G | Green | 11° Triple Bevel | 38 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15381537
|
Terumo™
8AN2125R1 |
21 G | Green | 11° Regular | 25 mm |
8.30 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15351537
|
Terumo™
8AN2025R1 |
20 G | Yellow | 11° Regular | 25 mm |
8.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15321537
|
Terumo™
8AN1838R1 |
18 G | Pink | 11° Regular | 38 mm |
8.55 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
- All needles are supplied sterile wrapped in individual, tamper proof peel pouches
- Simple color coded system allows easy identification of required sizes
- Use with a wide range of Sterile Syringes
- Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
- CE marked and latex free
- Supplied in boxes of 100 needles
Specifications
Sterile Hypodermic Needle for Single-Use | |
Sterile |
Safety and Handling
missing translation for 'shelfLife' : 5 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Terumo™ Agani™ Single-use Sterile Hypodermic Needles