missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Silencer™ GAPDH siRNA (human, mouse, rat)
Silencer™ GAPDH siRNA (human, mouse, rat)
305.00 € - 1432.00 €
Specifications
Control Type | Positive Control |
---|---|
RNAi Type | siRNA |
Species | Human, Mouse, Rat |
Label or Dye | Unconjugated |
Product Line | Silencer™, Ambion™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10772117
|
Invitrogen™
AM4624 |
5 nmol |
305.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10085804
|
Invitrogen™
AM4631 |
40 nmol |
740.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10371325
|
Invitrogen™
AM4632 |
5 x 40 nmol |
1432.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ambion™ Silencer™ GAPDH Positive Control siRNA (human, mouse, rat) is ideal for developing and optimizing siRNA experiments. The control is validated for use in human, mouse, and rat cell lines.- Validated siRNA controls for optimizing siRNA experiments
- Functionally tested in several common cell lines
- HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs.
Specifications
Positive Control | |
Human, Mouse, Rat | |
Silencer™, Ambion™ | |
HPLC |
siRNA | |
Unconjugated | |
siRNA | |
GAPDH |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Silencer™ GAPDH siRNA (human, mouse, rat)